Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145959.1 | NP_001139431.1 | 8, 9 | 750 | ||
NM_013864.2 | NP_038892.1 | 9, 10 | 792 | |||
GenBank mRNA | AB033921.1 | - | 9, 10 | 655 | ||
AK076514.1 | - | 6, 7 | 426 | |||
AK154597.1 | - | 9, 10 | 660 | |||
AK158900.1 | - | 8, 9 | 659 | |||
AK168011.1 | - | 8, 9 | 617 | |||
AK173136.1 | - | 6, 7 | 433 | |||
BC012963.1 | - | 9, 10 | 659 | 4211421 |