Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013865.2 | NP_038893.1 | 4 | 308 | ||
NM_180956.1 | NP_851287.1 | 4 | 308 | |||
GenBank mRNA | AB033922.1 | - | 4 | 222 | ||
AK002272.1 | - | 3 | 222 | |||
AK004689.1 | - | 4 | 254 | |||
AK036277.1 | - | 4 | 233 | |||
AK037487.1 | - | 4 | 234 | |||
AK154927.1 | - | 4 | 275 | |||
AK209466.1 | - | 1 | 22 | |||
BC018504.1 | - | 4 | 242 | 4500970 | ||
BY125191.1 | - | 4 | 309 |