Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166273.1 | NP_001159745.1 | 2 | 2469 | ||
NM_013884.3 | NP_038912.3 | 2 | 2469 | |||
GenBank mRNA | AF133700.1 | - | 2 | 654 | ||
AF461090.1 | - | 2 | 2463 | |||
AF461091.1 | - | 1 | 787 | |||
AF461092.1 | - | 2 | 2463 | |||
AK053891.1 | - | 1 | 51 | |||
AK138233.1 | - | 2 | 771 | |||
BC055736.1 | - | 2 | 801 | 6412504 |