Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024853.1 | NP_001020024.1 | 5, 6 | 474 | ||
NM_001024854.1 | NP_001020025.1 | 3, 4 | 290 | |||
NM_001286203.2 | NP_001273132.1 | 4, 5 | 376 | |||
NM_013896.3 | NP_038924.1 | 5, 6 | 512 | |||
GenBank mRNA | AF150101.1 | - | 4, 5 | 335 | ||
AK018764.1 | - | 3, 4 | 290 | |||
BC024370.1 | - | 5, 6 | 485 | 4976059 | ||
CK626183.1 | - | 3, 4 | 272 | |||
CO799389.1 | - | 4, 5 | 376 | |||
CO809703.1 | - | 443 | ||||
DV652207.1 | - | 5, 6 | 451 |