Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013908.4 | NP_038936.1 | 3, 4 | 476 | ||
GenBank mRNA | AF176520.1 | - | 3, 4 | 437 | ||
AK019715.2 | - | 3, 4 | 477 | |||
AK154911.1 | - | 2, 3 | 1415 | |||
AK218179.1 | - | 1, 2 | 121 | |||
BC010776.1 | - | 3, 4 | 403 | 4211592 |