Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002241.4 | NP_002232.2 | 2 | 407 | ||
GenBank mRNA | AK312383.1 | - | 2 | 314 | ||
BC034036.1 | - | 2 | 309 | |||
BC131627.1 | - | 2 | 303 | 40118533 | ||
DA958759.1 | - | 2 | 407 | |||
U52155.1 | - | 2 | 393 |