Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013925.4 | NP_038953.1 | 2, 3 | 328 | ||
GenBank mRNA | AF192375.1 | - | 1, 2 | 167 | ||
AK035614.1 | - | 2, 3 | 316 | |||
AK039699.1 | - | 2, 3 | 298 | |||
AK082399.1 | - | 2, 3 | 329 | |||
AK161044.1 | - | 2, 3 | 298 | |||
BC019976.1 | - | 2, 3 | 274 | 4458235 |