Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013930.4 | NP_038958.2 | 2, 3 | 339 | ||
GenBank mRNA | AJ224761.1 | - | 1, 2 | 230 | ||
AK044323.1 | - | 2, 3 | 320 | |||
AK098141.1 | - | 2, 3 | 280 | |||
AK136216.1 | - | 2, 3 | 288 | |||
AK149400.1 | - | 2, 3 | 264 | |||
BC005420.1 | - | 2, 3 | 325 | 3600654 | ||
BY275733.1 | - | 2, 3 | 327 | |||
CJ074697.1 | - | 2, 3 | 339 |