Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013933.3 | NP_038961.2 | 5 | 814 | ||
GenBank mRNA | AF115503.1 | - | 5 | 536 | ||
AF157497.1 | - | 5 | 755 | |||
AK003576.1 | - | 5 | 755 | |||
AK012357.1 | - | 5 | 569 | |||
AK149040.1 | - | 421 | ||||
AK159582.1 | - | 5 | 756 | |||
AK167381.1 | - | 5 | 815 | |||
AK168824.1 | - | 5 | 761 | |||
BC003866.1 | - | 5 | 800 | 3490082 |