Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289445.1 | NP_001276374.1 | 4 | 655 | ||
NM_001289446.1 | NP_001276375.1 | 4 | 385 | |||
NM_015731.3 | NP_056546.2 | 4 | 455 | |||
GenBank mRNA | AF152243.1 | - | 4 | 623 | ||
AK082271.1 | - | 4 | 381 | |||
AK089064.1 | - | 4 | 561 | |||
AK198418.1 | - | 3 | 251 | |||
AK201778.1 | - | 3 | 251 | |||
BC006949.1 | - | 2 | 157 | |||
BC021814.1 | - | 4 | 455 | |||
BC075718.1 | - | 4 | 583 | |||
BC094551.1 | - | 4 | 425 | 6813369 | ||
BY240030.1 | - | 4 | 386 |