Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130476.2 | NP_001123948.1 | 6 | 1781 | ||
NM_013837.2 | NP_038865.1 | 6 | 1598 | |||
GenBank mRNA | AF038008.1 | - | 6 | 1598 | ||
AK150753.1 | - | 6 | 1666 | |||
AK194543.1 | - | 1 | 125 | |||
BB805916.1 | - | 1 | 105 | |||
BC012666.1 | - | 6 | 1528 | 4035106 |