Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013843.3 | NP_038871.1 | 5 | 1591 | ||
GenBank mRNA | AB024004.1 | - | 1399 | |||
AK030965.1 | - | 5 | 1581 | |||
AK148857.1 | - | 5 | 1595 | |||
BC020362.1 | - | 5 | 1497 | |||
BC046328.1 | - | 5 | 1594 | 6475651 |