Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013851.2 | NP_038879.2 | 18 | 2590 | ||
GenBank mRNA | AF498362.1 | - | 18 | 2590 | ||
AK138196.1 | - | 18 | 2591 | |||
AK173034.1 | - | 17 | 2331 | |||
BC141401.1 | - | 18 | 2553 | 9056014 |