Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159284.1 | NP_001152756.1 | 18 | 2616 | ||
NM_001284427.1 | NP_001271356.1 | 17 | 2565 | |||
NM_001284428.1 | NP_001271357.1 | 18 | 2673 | |||
NM_001284429.1 | NP_001271358.1 | 18 | 2618 | |||
NM_013870.3 | NP_038898.2 | 17 | 2565 | |||
GenBank mRNA | AF064236.1 | - | 17 | 2512 | ||
AJ010305.1 | - | 17 | 2512 | |||
AK009426.1 | - | 5 | 978 | |||
AK030932.1 | - | 17 | 2566 | |||
AK083947.1 | - | 7 | 819 | |||
AK085412.1 | - | 18 | 2581 | |||
AK160492.1 | - | 8 | 1020 | |||
AK166385.1 | - | 18 | 2634 | |||
AK170516.1 | - | 17 | 2525 | |||
AK198309.1 | - | 1 | 113 | |||
BC010599.1 | - | 17 | 2506 | 3709078 | ||
BC066192.1 | - | 18 | 2491 | 30433616 | ||
BC069836.1 | - | 18 | 2491 | 30456568 | ||
BC069839.1 | - | 18 | 2491 | 30431675 |