Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013875.5 | NP_038903.3 | 13 | 4088 | ||
GenBank mRNA | AK035385.1 | - | 13 | 4101 |