Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033043.1 | NP_001028215.1 | 5 | 472 | ||
GenBank mRNA | AF190166.1 | - | 5 | 603 | ||
AF285585.1 | - | 5 | 472 | |||
AK136196.1 | - | 5 | 470 | |||
AY854010.1 | - | 5 | 472 | |||
AY854011.1 | - | 5 | 472 |