Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002250.2 | NP_002241.1 | 1 | 504 | ||
XM_005258882.2 | XP_005258939.1 | 1 | 1386 | |||
XM_005258883.2 | XP_005258940.1 | 1 | 1386 | |||
XM_011526938.1 | XP_011525240.1 | 1 | 1386 | |||
GenBank mRNA | AB128983.1 | - | 1 | 108 | ||
AB128984.1 | - | 1 | 108 | |||
AB128985.1 | - | 1 | 108 | |||
AF000972.1 | - | 1 | 108 | |||
AF022150.1 | - | 1 | 108 | |||
AF022797.1 | - | 1 | 504 | |||
AF033021.1 | - | 1 | 231 | |||
AF395661.1 | - | 1 | 179 | |||
BC015337.1 | - | 1 | 117 | 4431534 | ||
BT007426.1 | - | 1 | 108 |