Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013909.2 | NP_038937.2 | 9 | 1693 | ||
GenBank mRNA | AB041571.1 | - | 8 | 1931 | ||
AF176522.1 | - | 9 | 1681 | |||
AK040458.1 | - | 7 | 2073 | |||
AK052585.1 | - | 6 | 2106 | |||
BC011485.1 | - | 7 | 2066 | |||
BC026818.1 | - | 7 | 2060 | |||
BC044784.2 | - | 8 | 1893 | |||
BC054377.1 | - | 8 | 1514 | 6315438 |