Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013932.4 | NP_038960.2 | 10 | 1251 | ||
GenBank mRNA | AF142630.3 | - | 10 | 1182 | ||
AK050693.1 | - | 10 | 1246 | |||
AK078340.1 | - | 10 | 1210 | |||
BC024852.1 | - | 10 | 1195 | 5358404 | ||
BC061130.1 | - | 10 | 1206 | 6774579 |