Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289445.1 | NP_001276374.1 | 5 | 706 | ||
NM_001289446.1 | NP_001276375.1 | 5 | 436 | |||
NM_015731.3 | NP_056546.2 | 5 | 506 | |||
GenBank mRNA | AF011336.1 | - | 4 | 376 | ||
AF152243.1 | - | 5 | 674 | |||
AK082271.1 | - | 5 | 432 | |||
AK089064.1 | - | 5 | 612 | |||
AK185043.1 | - | 28 | ||||
BC006949.1 | - | 3 | 208 | |||
BC021814.1 | - | 5 | 506 | |||
BC075718.1 | - | 5 | 634 | |||
BC094551.1 | - | 5 | 476 | 6813369 | ||
BY240030.1 | - | 5 | 437 |