Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015732.4 | NP_056547.3 | 10, 11 | 2753 | ||
GenBank mRNA | AF073788.1 | - | 10, 11 | 2618 | ||
AF205889.1 | - | 9, 10 | 2458 | |||
AK049192.1 | - | 10, 11 | 2698 | |||
AK084644.1 | - | 3, 4 | 538 | |||
AK198476.1 | - | 2, 3 | 274 | |||
AK202759.1 | - | 2, 3 | 274 | |||
BC057338.1 | - | 10, 11 | 2736 | 6827741 |