Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015734.2 | NP_056549.2 | 65 | 5715 | ||
GenBank mRNA | AB009993.1 | - | 65 | 5455 | ||
AK148055.1 | - | 22 | 1859 | |||
AK157100.1 | - | 2 | 375 | |||
BC138447.1 | - | 65 | 5389 | 8861467 | ||
BC138449.1 | - | 65 | 5389 | 8861469 |