Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130412.1 | NP_001123884.1 | 5 | 579 | ||
NM_015763.4 | NP_056578.2 | 4 | 435 | |||
NM_172950.3 | NP_766538.2 | 4 | 435 | |||
GenBank mRNA | AK014526.1 | - | 3 | 396 | ||
AK019539.1 | - | 4 | 573 | |||
AK030921.1 | - | 4 | 438 | |||
AK141489.1 | - | 4 | 400 | |||
AK154837.1 | - | 5 | 576 |