Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010938.1 | NP_001010938.1 | 10, 11 | 1784 | ||
NM_001308046.1 | NP_001294975.1 | 10, 11 | 1842 | |||
NM_005781.4 | NP_005772.3 | 10, 11 | 1993 | |||
XM_005269268.3 | XP_005269325.1 | 10, 11 | 1784 | |||
XM_005269270.3 | XP_005269327.1 | 10, 11 | 1632 | |||
XM_005269274.3 | XP_005269331.1 | 7, 8 | 1049 | |||
XM_005269275.3 | XP_005269332.1 | 9, 10 | 1528 | |||
XM_011512317.2 | XP_011510619.1 | 12, 13 | 2351 | |||
XM_011512318.2 | XP_011510620.2 | 10, 11 | 1723 | |||
XM_011512320.1 | XP_011510622.1 | 10, 11 | 1506 | |||
XM_011512321.2 | XP_011510623.1 | 8, 9 | 1323 | |||
XM_017005508.1 | XP_016860997.1 | 10, 11 | 1723 | |||
XM_017005509.1 | XP_016860998.1 | 10, 11 | 1723 | |||
XM_017005510.1 | XP_016860999.1 | 10, 11 | 1723 | |||
GenBank mRNA | AB209338.1 | - | 7, 8 | 3851 | ||
AK127455.1 | - | 1, 2 | 106 | |||
AK131539.1 | - | 10, 11 | 1784 | |||
AK225786.1 | - | 10, 11 | 1789 | |||
BC020389.1 | - | 9, 10 | 1555 | |||
L13738.2 | - | 10, 11 | 2068 |