Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256796.1 | NP_001243725.1 | 3, 4 | 645 | ||
NM_001261397.1 | NP_001248326.1 | 2, 3 | 597 | |||
NM_001261398.1 | NP_001248327.1 | 2, 3 | 597 | |||
NM_002560.2 | NP_002551.2 | 2, 3 | 597 | |||
NR_046372.1 | 2, 3 | 597 | ||||
NR_046373.1 | 1, 2 | 449 | ||||
GenBank mRNA | AF000234.1 | - | 2, 3 | 193 | ||
AF012903.1 | - | 1, 2 | 79 | |||
AK225604.1 | - | 2, 3 | 293 | |||
AK293660.1 | - | 2, 3 | 329 | |||
AK297704.1 | - | 1, 2 | 181 | |||
AK301080.1 | - | 2, 3 | 281 | |||
AK308549.1 | - | 2, 3 | 329 | |||
AK308868.1 | - | 2, 3 | 329 | |||
BC017458.1 | - | 2, 3 | 326 | |||
BC033826.1 | - | 2, 3 | 317 | 5216449 | ||
BT019738.1 | - | 2, 3 | 293 | |||
BT019739.1 | - | 2, 3 | 293 | |||
BX402887.2 | - | 3, 4 | 417 | |||
U83993.1 | - | 2, 3 | 602 | |||
Y07684.1 | - | 2, 3 | 320 |