Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015775.2 | NP_056590.2 | 3 | 402 | ||
GenBank mRNA | AF113596.1 | - | 3 | 272 | ||
AF199362.1 | - | 3 | 283 | |||
AF243500.1 | - | 3 | 272 | |||
AK141350.1 | - | 3 | 376 | |||
AK146047.1 | - | 3 | 286 | |||
AK161173.1 | - | 3 | 396 | |||
BC038393.1 | - | 3 | 241 | 2648878 | ||
BC054348.1 | - | 3 | 402 | 2648889 |