Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015780.2 | NP_056595.1 | 1, 2 | 139 | ||
GenBank mRNA | BC111870.1 | - | 1, 2 | 122 | 40057075 | |
M29008.1 | - | 1, 2 | 139 |