Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015792.1 | NP_056607.1 | 3 | 266 | ||
GenBank mRNA | AK083118.1 | - | 3 | 611 | ||
AK147829.1 | - | 3 | 250 | |||
AK154357.1 | - | 3 | 289 | |||
AK158937.1 | - | 3 | 317 | |||
AK178185.1 | - | 2 | 117 | |||
AK178275.1 | - | 2 | 229 | |||
AK192093.1 | - | 2 | 143 | |||
AK204232.1 | - | 2 | 117 | |||
BC031393.1 | - | 3 | 266 | 4948804 |