Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122818.2 | NP_001116290.2 | 5, 6 | 480 | ||
NM_015801.2 | NP_056616.2 | 4, 5 | 421 | |||
GenBank mRNA | AF173829.1 | - | 4, 5 | 421 | ||
AK040720.1 | - | 5, 6 | 475 | |||
AK089095.1 | - | 2, 3 | 382 | |||
AK162641.1 | - | 2, 3 | 394 | |||
BC054789.1 | - | 5, 6 | 439 | 6831507 | ||
BC056999.1 | - | 5, 6 | 439 | 6831507 |