Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015811.2 | NP_056626.2 | 1, 2 | 200 | ||
GenBank mRNA | AF215667.1 | - | 1, 2 | 131 | ||
AK089293.1 | - | 1, 2 | 156 | |||
AK138688.1 | - | 1, 2 | 156 | |||
AK154394.1 | - | 156 | ||||
AK170366.1 | - | 1, 2 | 152 | |||
AK170957.1 | - | 1, 2 | 156 | |||
BC028634.1 | - | 1, 2 | 131 | 1328269 | ||
BU695313.1 | - | 1, 2 | 222 |