Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005452.1 | - | 3 | 366 | ||
AK049581.1 | - | 3 | 409 | |||
AK177565.1 | - | 2 | 115 | |||
AK179023.1 | - | 2 | 170 | |||
AK208995.1 | - | 2 | 115 | |||
AK214880.1 | - | 2 | 139 | |||
BC010332.1 | - | 3 | 387 | 2654393 | ||
BC016635.1 | - | 3 | 337 | |||
BC085104.1 | - | 3 | 325 | |||
BQ571983.1 | - | 2 | 212 | |||
CA540403.1 | - | 3 | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF123611.1 | - | 401 | Chr. - - [-] on Build |