Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015822.2 | NP_056637.1 | 2 | 481 | ||
GenBank mRNA | AF176521.1 | - | 2 | 188 | ||
AK080863.1 | - | 2 | 490 | |||
AK142007.1 | - | 2 | 481 | |||
AK166128.1 | - | 2 | 362 | |||
AK167409.1 | - | 2 | 300 | |||
BC026835.1 | - | 1 | 147 | |||
BC037665.1 | - | 1 | 144 | |||
BC046330.1 | - | 2 | 339 | |||
BC067203.1 | - | 2 | 363 | 6334317 |