Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088379.1 | - | 4, 5 | 422 | ||
BC010985.1 | - | 4, 5 | 425 | 4038194 | ||
CX239267.1 | - | 4, 5 | 418 | |||
M96265.1 | - | 4, 5 | 452 |