Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025427.3 | NP_001020598.1 | 3 | 368 | ||
NM_001039356.2 | NP_001034445.1 | 4 | 483 | |||
NM_001166535.1 | NP_001160007.1 | 3 | 406 | |||
NM_001166536.1 | NP_001160008.1 | 3 | 366 | |||
NM_001166537.1 | NP_001160009.1 | 3 | 660 | |||
NM_001166539.1 | NP_001160011.1 | 4 | 450 | |||
NM_001166540.1 | NP_001160012.1 | 3 | 373 | |||
NM_001166541.1 | NP_001160013.1 | 3 | 335 | |||
NM_001166542.1 | NP_001160014.1 | 3 | 333 | |||
NM_001166543.1 | NP_001160015.1 | 3 | 406 | |||
NM_001166544.1 | NP_001160016.1 | 3 | 368 | |||
NM_001166545.1 | NP_001160017.1 | 3 | 380 | |||
NM_001166546.1 | NP_001160018.1 | 3 | 368 | |||
NM_016660.3 | NP_057869.2 | 3 | 693 | |||
GenBank mRNA | AK010617.1 | - | 3 | 258 | ||
AK154957.1 | - | 3 | 289 | |||
AK155661.1 | - | 3 | 287 | |||
AK160036.1 | - | 3 | 257 | |||
AK169783.1 | - | 3 | 290 | |||
AK171334.1 | - | 3 | 322 | |||
AK182765.1 | - | 2 | 86 | |||
AK184734.1 | - | 2 | 86 | |||
AK185838.1 | - | 2 | 119 | |||
AK188296.1 | - | 2 | 119 | |||
AK199756.1 | - | 2 | 86 | |||
AK201985.1 | - | 2 | 119 | |||
BY270784.1 | - | 3 | 369 | |||
CF730372.1 | - | 3 | 596 | |||
CF951929.1 | - | 3 | 259 | |||
GH454882.1 | - | 4 | 445 | |||
J04179.1 | - | 3 | 362 |
Gene Symbol: Hmga1-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166476.1 | NP_001159948.1 | 1 | 289 | ||
NM_001166477.1 | NP_001159949.1 | 2 | 256 | |||
GenBank mRNA | BC008125.1 | - | 2 | 241 | ||
BC013455.1 | - | 1 | 299 | 4219805 | ||
BC093496.1 | - | 1 | 256 | 3601855 |