Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042966.1 | - | 2 | 142 | ||
BC107317.1 | - | 1 | 171 | |||
BC107318.1 | - | 1 | 171 | |||
BC119507.1 | - | 1 | 171 | |||
BC119508.1 | - | 1 | 171 | |||
BC127168.1 | - | 1 | 171 | |||
BC127169.1 | - | 1 | 171 | |||
U88401.1 | - | 1 | 170 |