Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016667.3 | NP_057876.1 | 1 | 666 | ||
GenBank mRNA | BC003748.1 | - | 1 | 439 | 3600695 | |
U89997.1 | - | 1 | 671 |