Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016670.3 | NP_057879.2 | 3, 4 | 349 | ||
GenBank mRNA | AF061270.1 | - | 3, 4 | 321 | ||
AK131788.1 | - | 2, 3 | 378 | |||
AK134000.1 | - | 4, 5 | 506 | |||
BC052701.1 | - | 2, 3 | 244 | 5721441 | ||
BY769139.1 | - | 349 |