Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016671.3 | NP_057880.2 | 3 | 274 | ||
GenBank mRNA | AF053005.1 | - | 3 | 242 | ||
AF265243.1 | - | 3 | 253 | |||
AK133753.1 | - | 3 | 289 | |||
BC032878.1 | - | 3 | 274 | 1244998 |