Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015773.2 | NP_056588.1 | 3 | 292 | ||
GenBank mRNA | AF173866.1 | - | 3 | 229 | ||
AK005732.1 | - | 3 | 240 | |||
AK054199.1 | - | 2 | 328 | |||
BC090660.1 | - | 3 | 235 | 30673350 | ||
BC100455.1 | - | 3 | 242 | 30919921 | ||
CJ060006.1 | - | 3 | 292 |