Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 2 | 414 | ||
NM_001285825.1 | NP_001272754.1 | 3 | 488 | |||
NM_015782.3 | NP_056597.3 | 3 | 491 | |||
GenBank mRNA | AK013886.1 | - | 3 | 465 | ||
AK088550.1 | - | 3 | 349 | |||
BC003229.1 | - | 3 | 391 | 3483391 | ||
BC094006.1 | - | 3 | 351 | 4983854 | ||
BC096648.1 | - | 3 | 345 | 1397390 | ||
CA552113.1 | - | 3 | 377 | |||
CX206945.1 | - | 3 | 491 | |||
CX565356.1 | - | 2 | 384 | |||
DV650448.1 | - | 1 | 107 | |||
L15447.1 | - | 3 | 287 |