Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015795.1 | NP_056610.1 | 4 | 289 | ||
GenBank mRNA | AF176531.1 | - | 4 | 289 | ||
AK043554.1 | - | 4 | 259 | |||
AK081947.1 | - | 4 | 335 | |||
AK164657.1 | - | 4 | 372 | |||
BC086690.1 | - | 4 | 253 | 30683387 | ||
BC115890.1 | - | 4 | 247 | 40103787 |