Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015807.1 | NP_056622.1 | 5 | 674 | ||
GenBank mRNA | AF078840.1 | - | 5 | 674 | ||
AK007418.1 | - | 5 | 713 | |||
BC024368.1 | - | 5 | 677 | 5065485 |