Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003740.3 | NP_003731.1 | 1 | 468 | ||
XM_005249456.1 | XP_005249513.1 | 1 | 201 | |||
GenBank mRNA | AB743589.1 | - | 1 | 104 | ||
AF084830.1 | - | 1 | 443 | |||
AK314298.1 | - | 1 | 201 | |||
BC060793.1 | - | 1 | 468 | 30348251 | ||
BC069573.1 | - | 1 | 175 | 7262361 | ||
EU978936.1 | - | 1 | 254 |