Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170745.1 | NP_001164216.1 | 3 | 799 | ||
NM_001170746.1 | NP_001164217.1 | 4 | 964 | |||
NM_015823.3 | NP_056638.1 | 3 | 799 | |||
GenBank mRNA | AK122349.1 | - | 2 | 259 | ||
AK147530.1 | - | 3 | 800 | |||
BC059005.1 | - | 3 | 456 | 6847238 |