Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003960.3 | NP_003951.3 | 2 | 792 | ||
GenBank mRNA | AB013094.1 | - | 2 | 793 | ||
AF187813.1 | - | 2 | 802 | |||
BC012626.1 | - | 2 | 780 | 4045439 | ||
BG426653.1 | - | 2 | 795 | |||
CR407610.1 | - | 1 | 625 |