Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088379.1 | - | 6 | 575 | ||
AK134525.1 | - | 3 | 228 | |||
AK163544.1 | - | 3 | 259 | |||
BB868849.1 | - | 3 | 286 | |||
BC010985.1 | - | 6 | 578 | 4038194 | ||
CX239267.1 | - | 6 | 571 | |||
M96265.1 | - | 6 | 605 |