Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016665.2 | NP_057874.2 | 4 | 221 | ||
GenBank mRNA | AK076181.1 | - | 3 | 280 | ||
AK080500.1 | - | 4 | 193 | |||
AK170126.1 | - | 4 | 193 | |||
BC052335.1 | - | 5 | 471 | |||
CA320983.1 | - | 3 | 160 | |||
U95003.1 | - | 5 | 285 | |||
U95005.1 | - | 5 | 459 |