Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015745.2 | NP_056560.1 | 4 | 3533 | ||
GenBank mRNA | BC017610.1 | - | 4 | 3533 | 4527589 | |
BC023069.1 | - | 4 | 824 | |||
BC034121.1 | - | 4 | 652 | |||
BC035273.1 | - | 4 | 1965 | |||
BC040282.1 | - | 4 | 3517 | |||
BC042480.1 | - | 4 | 3495 | 5363924 | ||
BC042509.1 | - | 4 | 726 | |||
BC083133.1 | - | 4 | 3552 | 6492729 |