Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCNKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042704.1 | NP_001036169.1 | 18, 19 | 2014 | ||
NM_001257139.1 | NP_001244068.1 | 17, 18 | 1888 | |||
NM_004070.3 | NP_004061.3 | 18, 19 | 2017 | |||
GenBank mRNA | AK225550.1 | - | 18, 19 | 1969 | ||
AK290814.1 | - | 18, 19 | 1972 | |||
AK298285.1 | - | 17, 18 | 1867 | |||
BC035373.1 | - | 22, 23 | 2541 | |||
BC048282.1 | - | 17, 18 | 1942 | 5184165 | ||
BC053869.1 | - | 19, 20 | 2062 | 5187202 | ||
Z30643.1 | - | 17, 18 | 1959 |