Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 6 | 1045 | ||
NM_001285825.1 | NP_001272754.1 | 7 | 1119 | |||
NM_015782.3 | NP_056597.3 | 7 | 1122 | |||
GenBank mRNA | AK013886.1 | - | 7 | 1096 | ||
AK088550.1 | - | 7 | 978 | |||
AK209909.1 | - | 2 | 218 | |||
AK210370.1 | - | 2 | 151 | |||
BC003229.1 | - | 7 | 1022 | 3483391 | ||
BC094006.1 | - | 7 | 982 | 4983854 | ||
BC096648.1 | - | 7 | 976 | 1397390 | ||
DV650448.1 | - | 5 | 738 | |||
L15447.1 | - | 7 | 918 |